Assay Details
| Gene Symbol: 5330425B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019908.1 | - | 1 | 852 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vascular cell adhesion molecule 1 | Vcam1 |
CD106, Vcam-1 |
-18.4kb | Anti-sense | Chr. 3 - 116110020 - 116129688 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 26544 |
Gm26544 | 1.3kb | Sense | Chr. 3 - 116150479 - 116165821 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5330425B07Rik
Genomic Map