Assay Details
| Gene Symbol: Taco1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019925.1 | - | 1 | 1353 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DDB1 and CUL4 associated factor 7 | Dcaf7 |
1700012F10Rik, 2610037L01Rik, C86529, HAN11, Wdr68 |
-5.8kb | Anti-sense | Chr. 11 - 106036872 - 106059324 on NCBI Build GRCm38 |
| Downstream | mitogen-activated protein kinase kinase kinase 3 | Map3k3 |
AW548911, MAPKKK3, Mekk3, mKIAA4031 |
18.1kb | Anti-sense | Chr. 11 - 106084902 - 106156656 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taco1os
Genomic Map