Assay Details
| Gene Symbol: 5330433J24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019930.1 | - | 1 | 1247 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CUB and Sushi multiple domains 3 | Csmd3 |
4930500N14Rik, mKIAA1894 |
-1.9mb | Sense | Chr. 15 - 47580633 - 48792231 on NCBI Build GRCm38 |
| Downstream | microRNA 1907 | Mir1907 |
Mirn1907, mmu-mir-1907 |
146.7kb | Sense | Chr. 15 - 50889025 - 50889114 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5330433J24Rik
Genomic Map