Assay Details
| Gene Symbol: 5430407F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019944.1 | - | 1 | 549 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HIV-1 tat interactive protein 2, homolog (human) |
Htatip2 |
AW111545, Cc3, Tip30 |
-17.6kb | Sense | Chr. 7 - 49759045 - 49778364 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 6 (neurotransmitter transporter, glycine), member 5 |
Slc6a5 |
Glyt2, prestin |
99.0kb | Sense | Chr. 7 - 49895702 - 49963864 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430407F15Rik
Genomic Map