Assay Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020024.1 | - | 1 | 753 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerisation domain containing 20 | Kctd20 |
2410004N11Rik, AI451943, AW541186, D17Ertd562e |
-37.5kb | Anti-sense | Chr. 17 - 28952448 - 28969549 on NCBI Build GRCm38 |
| Downstream | serine/arginine-rich splicing factor 3 | Srsf3 |
AL024116, Sfrs3, X16 |
24.8kb | Anti-sense | Chr. 17 - 29032660 - 29043372 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stk38
Genomic Map