Assay Details
| Gene Symbol: 6030400A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020045.1 | - | 1 | 1052 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 7854 | Gm7854 |
EG665934, ENSMUSG00000054483 |
-29.8kb | Anti-sense | Chr. 5 - 43151686 - 43235354 on NCBI Build GRCm38 |
| Downstream | C1q and tumor necrosis factor related protein 7 | C1qtnf7 |
5530401N20Rik, 8430425G24Rik, Ctrp7 |
249.0kb | Sense | Chr. 5 - 43515402 - 43619126 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6030400A10Rik
Genomic Map