Assay Details
| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020052.1 | - | 1 | 653 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, alpha 1C pseudogene |
Gm6682 | EG626534 | -34.0kb | Sense | Chr. 12 - 4782170 - 4783512 on NCBI Build GRCm38 |
| Downstream | FK506 binding protein 1b | Fkbp1b |
12.6kDa, AW494148, FKBP12.6, calstabin2 |
14.9kb | Anti-sense | Chr. 12 - 4833174 - 4842463 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sf3b6
Genomic Map