Assay Details
| Gene Symbol: 6720407P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020106.1 | - | 1 | 751 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibroblast growth factor 11 | Fgf11 |
Fgf-11, Fhf-3, Fhf3 |
Sense | Chr. 11 - 69796068 - 69802945 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 102 | Tmem102 |
4632403M07Rik, Cbap, Tmem102-ps |
660.0 | Sense | Chr. 11 - 69803595 - 69805624 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6720407P12Rik
Genomic Map