Assay Details
| Gene Symbol: 6720451E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020122.1 | - | 1 | 155 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurobeachin | Nbea |
Lyst2, mKIAA1544 |
Sense | Chr. 3 - 55625195 - 56183701 on NCBI Build GRCm38 | |
| Downstream | SEC13 homolog pseudogene | 4933417G07Rik | 12.6kb | Anti-sense | Chr. 3 - 55847105 - 55848065 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6720451E02Rik
Genomic Map