Assay Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020124.1 | - | 1 | 244 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitofusin 1 | Mfn1 |
2310002F04Rik, 6330416C07Rik, D3Ertd265e, HR2, mKIAA4032 |
-3.6kb | Anti-sense | Chr. 3 - 32529464 - 32579225 on NCBI Build GRCm38 |
| Downstream | actin-like 6A | Actl6a |
2810432C06Rik, AI851094, ARP4, Actl6, Baf53a, C79802 |
124.9kb | Anti-sense | Chr. 3 - 32708546 - 32726971 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gnb4
Genomic Map