Assay Details
| Gene Symbol: 6720454L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020125.1 | - | 1 | 151 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 378b | Mir378b |
mir-378b, mmu-mir-378b |
-178.7kb | Anti-sense | Chr. 11 - 88352773 - 88352864 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 0610039H22 gene | 0610039H22Rik | 79.4kb | Anti-sense | Chr. 11 - 88612086 - 88624512 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6720454L07Rik
Genomic Map