Assay Details
| Gene Symbol: 6720454L20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020126.1 | - | 1 | 849 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | brain protein I3 | Bri3 | I3 | Anti-sense | Chr. 5 - 144244437 - 144446757 on NCBI Build GRCm38 | |
| Downstream | BAI1-associated protein 2-like 1 | Baiap2l1 |
1300006M19Rik, AI585895, IRTKS |
16.8kb | Sense | Chr. 5 - 144264525 - 144358112 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6720454L20Rik
Genomic Map