Assay Details
| Gene Symbol: 6720484G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020175.1 | - | 1 | 451 | 168 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stromal antigen 1 | Stag1 |
AU045003, SA-1, Scc3 |
-51.9kb | Anti-sense | Chr. 9 - 100597709 - 100958544 on NCBI Build GRCm38 |
| Downstream | male-specific lethal 2 homolog (Drosophila) | Msl2 |
E130103E02Rik, Msl2l1, Rnf184 |
63.7kb | Anti-sense | Chr. 9 - 101100570 - 101104800 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6720484G13Rik
Genomic Map