Assay Details
| Gene Symbol: 8030425K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020195.1 | - | 1 | 650 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15412 | Gm15412 | OTTMUSG00000021245 | -243.8kb | Anti-sense | Chr. 7 - 96339483 - 96341964 on NCBI Build GRCm38 |
| Downstream | predicted gene 15413 | Gm15413 |
ENSMUSG00000053049, OTTMUSG00000021246 |
204.5kb | Anti-sense | Chr. 7 - 96791433 - 96801549 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8030425K09Rik
Genomic Map