Assay Details
| Gene Symbol: 8030488J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020221.1 | - | 1 | 553 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35147 |
Gm35147 | -108.1kb | Sense | Chr. 7 - 132460249 - 132466661 on NCBI Build GRCm38 | |
| Downstream |
NK1 transcription factor related, locus 2 (Drosophila) |
Nkx1-2 |
Nkx-1.1, Nkx-1.2, Sax1 |
20.5kb | Sense | Chr. 7 - 132596239 - 132599637 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8030488J09Rik
Genomic Map