Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020223.1 | - | 1 | 453 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | natriuretic peptide type C | Nppc |
CNP, lbab |
-186.8kb | Anti-sense | Chr. 1 - 86666293 - 86670573 on NCBI Build GRCm38 |
| Downstream | predicted gene 29371 | Gm29371 | 222.2kb | Sense | Chr. 1 - 87080262 - 87098594 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dis3l2
Genomic Map