Assay Details
| Gene Symbol: 9130231A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020290.1 | - | 1 | 748 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110051M20 gene | 1110051M20Rik | AI586322 | Sense | Chr. 2 - 91273240 - 91444713 on NCBI Build GRCm38 | |
| Downstream | low density lipoprotein receptor-related protein 4 | Lrp4 |
6430526J12Rik, D230026E03, Megf7, mdig |
128.6kb | Anti-sense | Chr. 2 - 91457470 - 91513901 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9130231A04Rik
Genomic Map