Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020355.1 | - | 1 | 750 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sperm motility kinase 3B | Smok3b |
4931427P12Rik, Smok2b2 |
-34.5kb | Sense | Chr. 5 - 138037224 - 138050633 on NCBI Build GRCm38 |
| Downstream | zinc finger and SCAN domain containing 21 | Zscan21 |
AI326272, CTfin51, RU49, Zfp-38, Zfp38, Zipro1 |
30.9kb | Sense | Chr. 5 - 138116899 - 138134265 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zkscan1
Genomic Map