Assay Details
| Gene Symbol: 9330199C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020396.1 | - | 1 | 748 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31456 |
Gm31456 | -603.0kb | Anti-sense | Chr. 14 - 38279215 - 38333385 on NCBI Build GRCm38 | |
| Downstream | predicted gene 20642 | Gm20642 | 284.2kb | Anti-sense | Chr. 14 - 39221626 - 39249746 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9330199C07Rik
Genomic Map