Assay Details
| Gene Symbol: 9430014K20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020417.1 | - | 1 | 853 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6377 | Gm6377 | EG622976 | -308.8kb | Sense | Chr. X - 109191425 - 109200448 on NCBI Build GRCm38 |
| Downstream | kinesin family member 18B pseudogene | Gm7134 | EG634340 | 862.6kb | Sense | Chr. X - 110372753 - 110375156 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430014K20Rik
Genomic Map