Assay Details
| Gene Symbol: 9430018C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020423.1 | - | 1 | 45 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calsyntenin 2 | Clstn2 |
2900042C18Rik, AI448973, CSTN2, Cs2, Cst-2, mKIAA4134 |
Sense | Chr. 9 - 97444395 - 98033384 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4921534H16 gene | 4921534H16Rik | 238.3kb | Anti-sense | Chr. 9 - 98003420 - 98012175 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430018C23Rik
Genomic Map