Assay Details
| Gene Symbol: 9430068D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020479.1 | - | 1 | 549 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane-associated ring finger (C3HC4) 11 | March11 | 9630025C22 | -235.0kb | Anti-sense | Chr. 15 - 26308904 - 26409594 on NCBI Build GRCm38 |
| Downstream | progressive ankylosis | Ank |
Ankh, D15Ertd221e, ank, mKIAA1581 |
821.4kb | Anti-sense | Chr. 15 - 27466677 - 27594909 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430068D22Rik
Genomic Map