Assay Details
| Gene Symbol: Malat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002847.2 | 1 | 3880 | 124 | ||
| GenBank mRNA | AK020483.1 | - | 1 | 644 | 124 | |
| EF177380.1 | - | 1 | 3880 | 124 | ||
| FJ209304.1 | - | 1 | 3880 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SCY1-like 1 (S. cerevisiae) | Scyl1 |
2810011O19Rik, C85140, Ntkl, mdf, mfd, p105 |
-27.0kb | Sense | Chr. 19 - 5758427 - 5771414 on NCBI Build GRCm38 |
| Downstream | nuclear paraspeckle assembly transcript 1 (non-protein coding) | Neat1 |
2310043N10Rik, VINC |
42.9kb | Sense | Chr. 19 - 5842302 - 5845478 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Malat1
Genomic Map