| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1446385_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1446385_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1446385_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1300012411.mm.1 |
NR_029463.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012411.mm.1 |
NR_029463.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012411.mm.1 |
TC1300001988.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300012407.mm.1 |
NR_029463.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012407.mm.1 |
NR_029463.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012407.mm.1 |
TC1300001988.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300012410.mm.1 |
NR_029463.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012410.mm.1 |
NR_029463.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012410.mm.1 |
TC1300001988.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300012409.mm.1 |
NR_029463.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012409.mm.1 |
NR_029463.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012409.mm.1 |
TC1300001988.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300012408.mm.1 |
NR_029463.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012408.mm.1 |
NR_029463.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300012408.mm.1 |
TC1300001988.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1446385_at |
- |
- |
| Mouse Expression Set 430 |
1446385_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1446385_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1446385_PM_at |
- |
- |