Assay Details
| Gene Symbol: 2300010F08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020509.1 | - | 1 | 548 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Eph receptor A7 | Epha7 |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
Sense | Chr. 4 - 28813044 - 28967511 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930556G01 gene | 4930556G01Rik | 1.7mb | Sense | Chr. 4 - 30664596 - 30855744 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2300010F08Rik
Genomic Map