Assay Details
| Gene Symbol: 9430087N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020513.1 | - | 1 | 247 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ER membrane protein complex subunit 2 | Emc2 |
4921531G14Rik, AV060620, AW209495, Ttc35 |
-205.8kb | Anti-sense | Chr. 15 - 43477214 - 43527777 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 74 | Tmem74 |
AA549547, B230382K22Rik |
132.0kb | Sense | Chr. 15 - 43866695 - 43870612 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430087N24Rik
Genomic Map