Assay Details
| Gene Symbol: 9530004M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020543.1 | - | 1 | 648 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase 7 | Map3k7 |
B430101B05, C87327, Tak1 |
Sense | Chr. 4 - 31962296 - 32023470 on NCBI Build GRCm38 | |
| Downstream | BTB and CNC homology 2 | Bach2 | E030004N02Rik | 229.4kb | Sense | Chr. 4 - 32238574 - 32586108 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530004M16Rik
Genomic Map