Assay Details
| Gene Symbol: 9530025L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020572.1 | - | 1 | 351 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ubiquitin-like, containing PHD and RING finger domains 2 |
Uhrf2 |
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf |
-71.0kb | Anti-sense | Chr. 19 - 30030433 - 30093729 on NCBI Build GRCm38 |
| Downstream | mannose-binding lectin (protein C) 2 | Mbl2 |
L-MBP, MBL, MBL-C, MBP-C |
67.1kb | Anti-sense | Chr. 19 - 30232906 - 30239686 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530025L08Rik
Genomic Map