Assay Details
| Gene Symbol: 9530027D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020577.1 | - | 1 | 651 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700006H21 gene | 1700006H21Rik | -159.8kb | Anti-sense | Chr. 13 - 107687397 - 107692168 on NCBI Build GRCm38 | |
| Downstream | uncharacterized LOC102634835 | LOC102634835 | 146.4kb | Anti-sense | Chr. 13 - 107999101 - 108004669 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530027D23Rik
Genomic Map