Assay Details
| Gene Symbol: 9530032E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020582.1 | - | 1 | 645 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 8 (sodium/calcium exchanger), member 1 |
Slc8a1 |
AI852629, AV344025, D930008O12Rik, Ncx1 |
Sense | Chr. 17 - 81373105 - 81738387 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 19689 |
Gm19689 | 1.4mb | Sense | Chr. 17 - 83033592 - 83078225 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530032E18Rik
Genomic Map