Assay Details
| Gene Symbol: 9530034D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020585.1 | - | 1 | 143 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
long intergenic non-protein coding RNA, Trp53 induced transcript |
Lincpint |
MNCb-1768, Pint, linc-Pint |
-112.2kb | Anti-sense | Chr. 6 - 31093563 - 31218474 on NCBI Build GRCm38 |
| Downstream |
muskelin 1, intracellular mediator containing kelch motifs, opposite strand |
Mkln1os |
Gm13845, OTTMUSG00000014733 |
35.2kb | Anti-sense | Chr. 6 - 31366885 - 31398773 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530034D02Rik
Genomic Map