Assay Details
| Gene Symbol: 9530067L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020630.1 | - | 1 | 945 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cirrhosis, autosomal recessive 1A (human) |
Cirh1a |
Cirhin, Naic, Teg-292, Tex292 |
-10.9kb | Anti-sense | Chr. 8 - 106893636 - 106923094 on NCBI Build GRCm38 |
| Downstream |
syntrophin, basic 2 |
Sntb2 | Snt2 | 712.0 | Anti-sense | Chr. 8 - 106935750 - 107014192 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530067L11Rik
Genomic Map