Assay Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020640.2 | - | 1 | 445 | 91 | |
| AK142313.1 | - | 1 | 1466 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700080O16 gene | 1700080O16Rik | -78.2kb | Sense | Chr. X - 51968693 - 51972793 on NCBI Build GRCm38 | |
| Downstream | glypican 3 | Gpc3 | OCI-5 | 220.8kb | Sense | Chr. X - 52272426 - 52614245 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpc4
Genomic Map