Assay Details
| Gene Symbol: 9630013K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020677.1 | - | 2 - 3 | 162 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1 | Hs3st3b1 |
3-OST-3B, 3OST3B1, 3Ost3b, AU041882, AW536289, Hs3st3b, m3-OST-3B |
-43.5kb | Anti-sense | Chr. 11 - 63884693 - 63922284 on NCBI Build GRCm38 |
| Downstream | predicted gene 12289 | Gm12289 | OTTMUSG00000005890 | 41.3kb | Sense | Chr. 11 - 64008145 - 64014488 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

9630013K17Rik
Genomic Map