Assay Details
| Gene Symbol: A330102K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020729.1 | - | 1 | 943 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
succinate-Coenzyme A ligase, GDP-forming, beta subunit |
Suclg2 |
AF171077, AW556404, D6Wsu120e, SCS-betaG |
-316.6kb | Sense | Chr. 6 - 95473009 - 95718846 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 19, member A1 |
Fam19a1 |
C630007B19Rik, FAM19A1 TAFA-1, TAFA1, Tafa-1 |
76.2kb | Anti-sense | Chr. 6 - 96112775 - 96657204 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A330102K18Rik
Genomic Map