Assay Details
| Gene Symbol: A430102A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020755.1 | - | 1 | 549 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 57 | Ccdc57 | 4933434G05Rik | -48.0 | Anti-sense | Chr. 11 - 120826529 - 120932902 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 16 (monocarboxylic acid transporters), member 3 |
Slc16a3 |
Mct3, Mct4 |
14.8kb | Sense | Chr. 11 - 120948484 - 120959000 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A430102A20Rik
Genomic Map