Assay Details
| Gene Symbol: A430105P17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020769.1 | - | 1 | 852 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
POU domain class 5, transcription factor 2 |
Pou5f2 |
1700013G10Rik, SPRM-1, Sprm1 |
-7.2kb | Sense | Chr. 13 - 78024902 - 78026296 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 38604 |
Gm38604 | 90.1kb | Anti-sense | Chr. 13 - 78124863 - 78183977 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A430105P17Rik
Genomic Map