| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300000044.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000044.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000044.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000047.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000047.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000047.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000051.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000051.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000051.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000050.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000050.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000050.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000052.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000052.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000052.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000045.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000045.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000045.mm.1 |
TC1300000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300000046.mm.1 |
NR_045363.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000046.mm.1 |
NR_045363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300000046.mm.1 |
TC1300000005.mm.1 |
MTA TC |