Assay Details
| Gene Symbol: C030007I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI853428.1 | - | 1 | 105 | ||
| AK021055.1 | - | 1 | 50 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calneuron 1 | Caln1 |
9630012C17Rik, Cabp8, MNCb-0849 |
Sense | Chr. 5 - 130369356 - 130846325 on NCBI Build GRCm38 | |
| Downstream | Williams-Beuren syndrome chromosome region 17 homolog (human) | Wbscr17 |
AA388321, E330012B09Rik |
30.1kb | Anti-sense | Chr. 5 - 130874951 - 131307522 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030007I01Rik
Genomic Map