Assay Details
| Gene Symbol: Gm3985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177589.1 | NP_001171060.1 | 2 - 3 | 247 | 76 | |
| GenBank mRNA | AK158352.1 | - | 2 - 3 | 248 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuregulin 1 | Nrg1 |
6030402G23Rik, ARIA, D230005F13Rik, GGF, GGFII, HRG, HRGalpha, Hgl, NDF, SMDF |
-39.4kb | Sense | Chr. 8 - 31807452 - 32890583 on NCBI Build GRCm38 |
| Downstream | Werner syndrome homolog (human) | Wrn | AI846146 | 291.9kb | Sense | Chr. 8 - 33234372 - 33385545 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158352.1, NM_001177589.1 transcripts.
Set Membership: |
|

Gm3985
Genomic Map