Assay Details
| Gene Symbol: Olfr1077-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF178753.1 | - | 1 | 156 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactory receptor 1076 | Olfr1076 | MOR189-2 | -16.0kb | Anti-sense | Chr. 2 - 86508461 - 86509402 on NCBI Build GRCm38 |
| Downstream | olfactory receptor 1079 | Olfr1079 | MOR189-1 | 12.1kb | Sense | Chr. 2 - 86537966 - 86542480 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1077-ps1
Genomic Map