Assay Details
| Gene Symbol: Gm9777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019581.1 | - | 2 | 247 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nicotinamide nucleotide transhydrogenase | Nnt |
4930423F13Rik, AI323702, BB168308 |
Anti-sense | Chr. 13 - 119335527 - 119409138 on NCBI Build GRCm38 | |
| Downstream | polyadenylate binding protein-interacting protein 1 | Paip1 | PAIP-1 | 18.8kb | Sense | Chr. 13 - 119428271 - 119460323 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9777
Genomic Map