Assay Details
| Gene Symbol: Gm41043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029426.1 | - | 1 - 2 | 200 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4922502H24 gene | 4922502H24Rik | -146.2kb | Sense | Chr. 13 - 101391871 - 101397580 on NCBI Build GRCm38 | |
| Downstream |
phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Pik3r1 |
PI3K, p50alpha, p55alpha, p85alpha |
132.6kb | Sense | Chr. 13 - 101680761 - 101768217 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm41043
Genomic Map