Assay Details
| Gene Symbol: Gm28590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031624.1 | - | 3 - 4 | 322 | 161 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 18 | Ddx18 | 2310005B10Rik | -664.1kb | Anti-sense | Chr. 1 - 121553830 - 121567980 on NCBI Build GRCm38 |
| Downstream | dipeptidylpeptidase 10 | Dpp10 |
6430601K09Rik, DPP X, Dprp3 |
1.1mb | Anti-sense | Chr. 1 - 123332138 - 124845960 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm28590
Genomic Map