Assay Details
| Gene Symbol: Gm28548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033064.1 | - | 2 - 3 | 241 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
acid phosphatase, prostate |
Acpp |
5'-NT, A030005E02Rik, FRAP, Lap, PAP, Ppal |
-44.6kb | Anti-sense | Chr. 9 - 104288240 - 104337722 on NCBI Build GRCm38 |
| Downstream | microRNA 2136 | Mir2136 | mmu-mir-2136 | 41.1kb | Sense | Chr. 9 - 104426113 - 104426187 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm28548
Genomic Map