Assay Details
| Gene Symbol: Gm11269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132790.1 | - | 1 - 2 | 46 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11267 | Gm11267 | OTTMUSG00000000407 | -102.2kb | Anti-sense | Chr. 4 - 82584573 - 82590609 on NCBI Build GRCm38 |
| Downstream |
zinc finger, DHHC domain containing 21 |
Zdhhc21 |
9130404H11Rik, AL024349, D130004H04Rik, dep |
105.7kb | Anti-sense | Chr. 4 - 82798738 - 82859891 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11269
Genomic Map