Assay Details
| Gene Symbol: Gm36184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132862.1 | - | 1 - 2 | 637 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and MYND domain containing 1 | Ankmy1 |
4930483I10Rik, ZMYND13 |
Anti-sense | Chr. 1 - 92859844 - 92902986 on NCBI Build GRCm38 | |
| Downstream | dual specificity phosphatase 28 | Dusp28 |
0710001B24Rik, AV005521 |
39.7kb | Sense | Chr. 1 - 92906989 - 92908620 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm36184
Genomic Map