Assay Details
| Gene Symbol: Gm30807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132883.1 | - | 1 - 2 | 170 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GSG1-like | Gsg1l |
AI852434, C230098I05Rik, G630023A01Rik, Gm637, g1l |
-9.2kb | Sense | Chr. 7 - 125878412 - 126082772 on NCBI Build GRCm38 |
| Downstream | exportin 6 | Xpo6 |
2610005L19Rik, AI854665, AL022631, C230091E20Rik, R75304, Ranbp20, mKIAA0370 |
8.4kb | Sense | Chr. 7 - 126101718 - 126200496 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm30807
Genomic Map