Assay Details
| Gene Symbol: Gm15509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133047.1 | - | 1 - 2 | 1885 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 1, Pasteur Institute 1 |
D1Pas1 | Pl10 | -151.3kb | Anti-sense | Chr. 1 - 186967416 - 186970627 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700007P06 gene | 1700007P06Rik | 773.0 | Anti-sense | Chr. 1 - 187125138 - 187127852 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15509
Genomic Map