Assay Details
| Gene Symbol: Gm31456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133121.1 | - | 811 | 159 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth hormone inducible transmembrane protein | Ghitm |
1010001P14Rik, C77840, MICS1, PTD010, Tmbim5 |
-1.2mb | Anti-sense | Chr. 14 - 37120444 - 37135327 on NCBI Build GRCm38 |
| Downstream | neuregulin 3 | Nrg3 | ska | 31.7kb | Anti-sense | Chr. 14 - 38365102 - 39473113 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31456
Genomic Map